Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix NATtrol™ Influenza External Run Controls
Formulated with purified, intact chemically modified virus particles to render them non-infectious and refrigerator stable
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Type | Influenza External Run Control |
For Use With (Application) | Quality control of clinical assays and training of laboratory personnel |
Quantity | 6 x 0.5 mL |
Catalog Number | Mfr. No. | Detectable Analytes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Detectable Analytes | Price | Quantity & Availability | |||||
22-156-723
|
ZeptoMetrix
NATCXVA96MC |
Coxsackie virus A9 Nucleic Acid |
Each for $303.00
|
N/A | |||||
22-156-730
|
ZeptoMetrix
NATFLUA/B6MC |
Influenza A/NewCaledonia/20/99 Nucleic Acid, Influenza B/Florida/02/07 Nucleic Acid |
Each for $296.00
|
N/A | |||||
22-156-733
|
ZeptoMetrix
NATFLUAH1N16MC |
Influenza A/NY/02/09 Nucleic Acid |
Each for $296.00
|
N/A | |||||
Description
- Each control contains 6 x 0.5mL vials of NATtrol™ Influenza A/B or Influenza A H1N1 (2009) or Coxsackie virus A9
- Supplied in purified protein matrix identical to the composition of true clinical specimen
- Designed to evaluate the performance of nucleic acid tests for determination of presence of respiratory virus nucleic acids
Contain intact organisms and should be run in a manner identical to that used for clinical specimens.
Quality control of clinical assays and training of laboratory personnel
Specifications
2°C to 8°C | |
Influenza External Run Control | |
Quality control of clinical assays and training of laboratory personnel | |
6 x 0.5 mL |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title