Learn More
ZeptoMetrix Immuno-Tek Human IgM ELISA
Measures human IgM in serum, plasma, hybridoma cell supernatants, ascites or other biological fluids
Supplier: ZeptoMetrix 0801183
Includes: One 96 well microplate, 12mL detector antibody, 7mL of Human IgM standard, 100mL assay diluent, 125mL of plate wash buffer, 12mL substrate, 12mL stop solution, 1 pack of microplate sealers , 1 plastic bag.
Description
- Useful in monitoring production and purification of human monoclonal antibodies
- Contains ready–to–use reagents
- Takes less than two hours to perform
Specifications
Immuno-Tek Human IgM ELISA kit | |
Designed for the measurement of Human IgM in Serum, Plasma, hybridoma cell supernatants, ascites or other biological fluids, the kit is especially useful in monito ring the production and purification of Human Monoclonal Antibodies | |
96 Determinations |
96-well Microplate | |
1 96-well microplate, 1 pack of microplate sealers, 1 plastic bag, 100 mL assay diluent, 12 mL detector antibody, 12 mL stop solution, 12 mL substrate, 125 mL plate wash buffer, 7 mL of Human IgM standard | |
2°C to 8°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For research use only. Not for in vitro diagnostic use.