Learn More
Acs Material ZEOLTC IMDZLT FRMWRK-67, 5GR
SDP
Supplier:  Acs Material MMZ67B05
Encompass
Zeolitic Imidazolate Framework-67 (ZIF-67), 5 GR.Type B: Solvent Thermal Method + Dried at 150 ̊C for 8 hrs
Catalog No. NC2108285