Learn More
Thermo Scientific Chemicals Zeocin^t, 100 mg/ml in HEPES, sterile-filtered
Zeocin (phleomycin, CAS # 11031-11-1) is a broad-spectrum antibiotic that acts by intercalating DNA.
$80.90 - $278.00
Specifications
Chemical Name or Material | Zeocin™ |
---|---|
MDL Number | MFCD28100499 |
Solubility Information | Fully miscible. |
Physical Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AAJ67140XF
|
Thermo Scientific Chemicals
J67140XF |
1 mL |
Each for $80.90
Order in Bulk
Request bulk or custom formats |
|
|||||
AAJ671408EQ
|
Thermo Scientific Chemicals
J671408EQ |
5 x 1 mL |
Pack for $278.00
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Zeocin, a popular and effective antibiotic for the selection of vectors bearing the Sh ble gene in a variety of cells types (bacteria, eukaryotic microorganisms, plant and animal cells). Resistance to Zeocin is conferred by the Sh ble gene product, which inactivates Zeocin™ by binding to the antibiotic. Sh ble is a small gene, only 370bp in size, carried in a number of InvivoGen’s vectors. Zeocin is used as a selective agent in molecular genetics experiments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Zeocin (phleomycin) is a glycopeptide from the bleomycin family of antibiotics
- Zeocin intercalates DNA, producing double-stranded breaks and preventing cell division
- It can chelate some active metals including cobalt, iron, and copper
- Zeocin has antineoplastic and antifungal properties
Application
- Zeocin is useful for genetic selection experiments
Specifications
Zeocin™ | |
MFCD28100499 | |
Fully miscible. | |
Liquid |
Safety and Handling
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.