Learn More
PIP™ Zenon Z12R Bi-Focals
Click to view available options
Frame Material:
Nylon
Polycarbonate
Magnification Power:
+1.00
+1.25
+1.50
+1.75
+2.00
+2.25
+2.50
+2.75
+3.00
Description
- Lightweight, rimless design for user comfort
- Flexible, rubber tip temples provide a secure fit without uncomfortable pressure points
- Variety of lenses to fit applications
- Molded nose bridge design to fit the vast majority of users
Specifications
Lens Coating | Scratch-resistant |
Lens Tint | Clear |
Frame Color | Clear |
Magnification Power | +3.00 |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |
UV Protection | 0.9999 |
Warning - Prop 65 - Cancer and Reproductive Harm