Learn More
Electron Microscopy Sciences Bio-Pump Plus
Supplier:  Electron Microscopy Sciences 6521112
Encompass_Preferred
The Zefon Bio-Pump Plus is the smallest and lightest portable battery powered IAD sampling pump available. It provides the simplest and most convenient way to sample with Air-O-Cell and Via-Cell cassettes at a flow rate of 15 LBM.
New "quick sample" feature makes sampling a breeze.
Extra long batter life allowing for over 100 samples in a single charge.
Quick-charge battery charger included (approximately 3 hours).
Programmable timer allows unattended operation.
Nothing to prepare or clean-just place the cassette on the pump, set the time and go.
Complete with both an Air-O-Cell and Via-Cell flow indicator, quick charger, 10 free Air-O-Cell cassettes and a handy carrying case.
Catalog No. 50-338-82