Learn More
Paul N Gardner Company ZAHN PAINT VISCOMETER CUP NO2
Supplier: Paul N Gardner Company VIEZ2
CALIBRATED EZ ZAHN (ASTM) DIP VISCOSITY CUPS, No. 2 EZ Dip Viscosity Cup with Conversion Table, SECONDS RANGE :20 to 60 , CENTISTOKE SENSITIVITY :19 to 156 , MIDRANGE OIL NUMBER:3.3