Learn More
Enzo Life Sciences Z-Pro-Leu-Gly-NHOH (100mg)
Supplier: Enzo Life Sciences BMLPI1390100
A short peptide hydroxamate that inhibits MMPs, astacin, meprin a, and likely other metzincin family members. Can also be used for purification of collagenase. Purity: ≥98%. Long Term Storage: -20°C.