Learn More
Biotium Z-FR-R110, 5MG
Supplier:  Biotium 10209
Encompass_Preferred
Z-FR-R110 (Rhodamine 110, bis-(N-CBZ-L-phenylalanyl-L-arginine amide), dihydrochloride) is a substrate for the cysteine proteases cathepsins B and L. The substrate has been used in flow cytometry studies of human monocytes and rat macrophages.
Catalog No. NC2271078