Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HORIBA Yumizen C1200 Cystatin C Control
Yumizen C1200 Cystatin C Control used to monitor Cystatin C assay
Specifications
Certifications/Compliance | CE Marked |
---|---|
Content And Storage | 2°C to 8°C |
Detectable Analytes | Cystatin C |
For Use With (Equipment) | Used on Yumizen C1200 Clinical Chemistry Analyzer |
Sample Type | Serum |
Catalog Number | Mfr. No. | Concentration | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Concentration | Price | Quantity & Availability | |||||
23-601-597
|
HORIBA
1300023951 |
Normal concentration |
Each of 1 for $289.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Used to monitor Cystatin C assay
Specifications
CE Marked | |
Cystatin C | |
Serum | |
Control | |
3 Pk. 1 mL Vials | |
12 weeks (at 2°C to 8°C) | |
Liquid |
2°C to 8°C | |
Used on Yumizen C1200 Clinical Chemistry Analyzer | |
Assay Control | |
Single-use | |
Recombinant Cystatin C | |
Non-sterile |
Safety and Handling
ShelfLife : 9 months
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title