Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YSI™ TruLine pH Combination Electrode with Gel Reference Electrolyte, pH 26
Single fiber junction, 3-in-1 benchtop electrode.
Supplier: YSI™ 400352
Includes: BNC connector, 1m cable, one banana plug
Description
- For use with TruLab Instruments
- Measures pH and temperature
- Epoxy shaft
- Buffer recognition
- User calibratable and user replaceable
Specifications
- pH Range: 0 to 14
- Operating Temperature: -5° to +80°C (23° to 176°F)
Order Info
Item is heat packed in the winter months. Shipment must be made via 2-day or next day carrier during that time.
Compliance
GLP compliant
Warranty and Services
One year
Specifications
TruLine pH 26 Combination Electrode | |
1m cable, BNC connector, Banana plug | |
0 to 14 | |
TruLab Instruments |
Measuring pH and temperature | |
Epoxy | |
-5°C to +80°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction