Learn More
YSI™ Starter Nickel Reagent Test Kits for EcoSense 9300/9500 Photometer
Nickel test kit for use with for EcoSense 9300 or 9500 Photometers. Eliminate sample preservation by taking your portable, waterproof instruments to the field to perform the analysis on-site. All reagents are safe and easy to use.
Supplier: YSI™ YPM284
Includes: Reagents for 50 tests, instructions and accessories
Description
- 0 to 10.0 range
- 50 count
- Not suitable for seawater use; sample may be diluted to lower salt content to help avoid precipitate that can interfere with testing; results may vary
Specifications
Nickel Reagent Test Kit | |
EcoSense 9300/9500 Photometer | |
Reagents for 50 tests, instructions and accessories |