Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YSI™ Science pH Combination Electrode with Silamid™ Double Junction, pHT-Pt pH
3-in-1, Platinum junction benchtop electrode.
Supplier: YSI™ 400360
Description
- For use with TruLab Instruments
- Glass shaft
- Measures pH and temperature
- BNC connector and one banana plug
- Buffer recognition
- User calibratable and replaceable
- Refillable
- Sphere (bulb) membrane
- Reference Electrolyte: 3M KCl liquid
Specifications
- pH Range: 0 to 14
- Temperature Range: -5° to +100°C (23° to 212°F)
- NTC: 30 kOhm
- Length: 170mm
- Cable Length: 1m
- Dia.: 12mm
- Min. Immersion Depth: 17 to 19mm
Order Info
Item is heat packed in the winter months. Shipment must be made via 2-day or next day carrier during that time.
Compliance
GLP compliant
Warranty and Services
One year
Specifications
Science pHT-Pt pH Combination Electrode | |
1m cable, BNC connector, one banana plug | |
0 to 14 | |
TruLab Instruments |
Measuring pH and temperature | |
Glass | |
-5°C to +100°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction