Learn More
Med Vet International Boston Round Yorker Dropper Bottle, 60ml/2oz, 100/Case
Supplier: Med Vet International BSRD60CS
Removeable dropper tips allow refilling. Bottle and tips are made of low density polythelene. Screw cap made of polystyrene.