Learn More
MSC SPI CMS160725026 0" to 6" Range, 0.001" Graduation, 0.1" per Revolution, Dial Caliper White Face, 1.57" Jaw Length, Accurate to 0.0010", NIST Traceability Certificate
Supplier: MSC 37530623
SPI CMS160725026 0" to 6" Range, 0.001" Graduation, 0.1" per Revolution, Dial Caliper White Face, 1.57" Jaw Length, Accurate to 0.0010", NIST Traceability Certificate