Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nasco XR Clinic Mobile - Virtual Reality Medical Training Solution
XR Clinic Mobile - Virtual Reality Medical Training Solution is a new way to develop skills using virtual patients.
Supplier: Nasco XRCM010
Description
- A revolutionary way to develop skills with high resolution graphics.
- Virtual patients answer questions allowing learners to develop their medical diagnostic abilities
- Industry leading graphics
- Minimum space needed for use: 4 sq. ft.
- Lifetime license
- Unique teleportation feature inside VR section
- Fastest setup, load and response time
- 200+ medical scenarios in library
- Case Manager for creating and editing your own cases
- Smart Center with Spectator feature for managing and tracking training
- Multi-user capability – ideal for medical schools or simulation centers
- Multiple languages: English, Spanish, Arabic, Turkish, Portuguese, German, and more to come
- Dimensions: 36 cm L x 22 cm W x 13.5 cm H
- Ship weight: 2.3 kg (5.07 lb.)
Specifications
VR Mobile Clinic | |
Medical Training | |
White | |
36 cm x 22 cm x 13.5 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction