Learn More
Enzo Life Sciences Xestospongin C (50µg). CAS: 88903-69-9
Supplier: Enzo Life Sciences BMLCA4090050
Xestonspongin C is a structurally novel marine alkaloid isolated from the Okinawan sponge Xestospongia sp. It is a potent, cell-permeable inhibitor of IP3 receptor-mediated Ca2+ release (IC50 = 358 nM). However, since xestospongin C also inhibits voltage-dependent Ca2+ and K+ currents at concentrations similar to those which inhibit the IP3 receptor, it can only be regarded as a selective blocker of the IP3 receptor in permeabilized cells and not in cells with intact plasma membranes. Has vasodilatory properties. Purity: ≥90% (TLC). Solubility: Soluble in DMSO or 100% ethanol at 2mM. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.