Learn More
Strem, An Ascensus Company Xenon(II) fluoride, 99.5% 2g
SDP
Supplier:  Strem, An Ascensus Company 5415002G
Encompass_Preferred
CAS# 13709-36-9. 2g. Xenon(II) fluoride, 99.5%. MFCD00040538. Molecular Weight: 169.30. Molecular Formula: XeF2. Color/form: white xtl. Strem# 54-1500. http://www.strem.com/catalog/v/54-1500/
Catalog No. 50-901-12658
Explore available promotions