Learn More
Spectra Services Inc X-CITE 120 SERIES REPL LAMP
Supplier: Spectra Services Inc 01263000
The powerful 120W lamp used in the X-Cite 120Q Series provides excitation intensity comparable to, or better than, conventional mercury arc lamps, with an added benefit of a broader spectrum being able to excite more fluorophores. The lamps have a guaranteed life of 2000 hours.