Learn More
Microtech Scientific WTR SMPL BTL120ML PS100BX/2/CS
Supplier: Microtech Scientific MS5RT120PS
Water Sample Bottles, 120mL Polystyrene, with Sodium Thiosulfate and Paper Tamper Seal, Sterile, 100/Box, 200/Case