Learn More
Med Vet International Cloth Instrument Wrap, 36" x 36", Each
Supplier: Med Vet International W3636
Wrappers are a woven blend of 55% cotton and 45% polyesterMedium weight, 4.1 oz. per square yardThe T-160 thread count and fabric weight ensures durabilityThese two-ply wrappers are stitched with a bias binding that is also color-coded for quick identification