Learn More
Med Vet International Cloth Instrument Wrap, 18" x 18", Each
Supplier: Med Vet International W1818
Wrap are a woven blend of 55% cotton and 45% polyester Medium weight, 4.1 oz. per square yard The T-160 thread count and fabric weight ensures durability This two-ply wrap is stitched with a bias binding that is also color-coded for quick identification