Learn More
Junkin Safety Appliance Wrap Around Fire Blanket Kit
SDP
Supplier:  Junkin Safety Appliance JSA1010
Encompass_Preferred
Kit consists of Fire Blanket (JSA-1010-B) Fire Resistant Treated 100% Wool for maximum fire protection and sturdy 20 Ga. Steel Cabinet (JSA-1010-CW) ready for wall mounting. Cabinet is bright red enamel with white lettering. Viewing hole provided to see if blanket is in place. The Wrap Around Fire Blanket Kit is designed so a person can insert either arm through the rope loop and, by rotating their body, completely wrap themselves in the fire blanket. Kit should be wall mounted 6" to 8" from floor. Complete kit meets MIL-B-52309B.
Catalog No. NC0634861