Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Wood Handled Cotton Swabs and Applicators
Useful in bacteriology labs for inoculating broths or plated media or for obtaining culture specimens, and for cleaning delicate instrument.
Description
- Multi-purpose applicators ideal for specimen collection and use in a wide range of diagnostic testing and screening
- Sterile and non-sterile products available (see product specifications)
- Polyester-tipped and cotton applicators on wooden or plastic handles available (see product specifications)
- Not suitable for PCR DNA/ RNA testing
Specifications
Cotton Tip Applicator | |
Wood | |
1/ envelope; 1000/cs | |
15 cm | |
Useful in bacteriology labs for inoculating broths or plated media or for obtaining culture specimens and for cleaning delicate instruments |
Sterilized by Gamma Radiation | |
Cotton | |
Sterile | |
6 in. |
missing translation for 'provideContentCorrection'
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'