Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSC Go Science Crazy Wire Gauze

Aids in the distribution of heat while using a Bunsen burner. GSC International Wire Gauze features rolled smooth wire edges. A variety of sizes are available.
$8.05 - $10.50
Specifications
Description | Rolled smooth wire edges |
---|---|
Material | Wire |
Alert:
Intended for ages 13 and up.
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
S13736
![]() |
GSC Go Science Crazy
4000612 |
6 x 6 in. |
Pack of 12 for $10.50
|
|
S13735
![]() |
GSC Go Science Crazy
4000512 |
5 x 5 in. |
Pack of 12 for $9.20
|
|
S13734
![]() |
GSC Go Science Crazy
4000412 |
4 x 4 in. |
Pack of 12 for $8.05
|
|
Description
- Wire gauze is used with Bunsen burners to aid in the distribution of heat
- Helps protect lab equipment
- Features rolled smooth wire edges
- Available in several sizes
Specifications
Rolled smooth wire edges | |
Wire |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title