Learn More
MSC Value Collection K40040000101212 0.0011 Inch Wire Diameter, 400 x 400 Mesh per Linear Inch, Stainless Steel, Wire Cloth 0.0015 Inch Opening Width, 12 Inch Wide x 12 Inch Stock Length
Supplier:  MSC 52431459
Encompass
Value Collection K40040000101212 0.0011 Inch Wire Diameter, 400 x 400 Mesh per Linear Inch, Stainless Steel, Wire Cloth 0.0015 Inch Opening Width, 12 Inch Wide x 12 Inch Stock Length
Catalog No. NC1606322