Learn More
Chemplex Industries WINDOW FRAME 0.24 MIL 3.0IN DI
Supplier:  Chemplex Industries 3020
Encompass_Preferred
CHEMPLEX SPECTROMEMBRANE(TM) Thin-Film Sample Support Window Frames, Polypropylene, 6.0 u (0.24 mil), 3.0 Dia., In.
Catalog No. 50-249-359