Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Wilmad™ Thin Wall Quartz EPR Sample Tube

Manufactured from naturally occurring quartz with the highest purity to minimize the background signal. Bel-Art SP Scienceware™ Thin Wall Quartz EPR Sample Tube may contain Fe2O3 which is paramagnetic (1200 ppm) and has certain impact on shimming quality depending on the magnetic field strength.

Supplier:  Bel-Art™ 660000015

 View more versions of this product

Catalog No. 16-800-115


Add to cart

Description

Description

  • Tight ID and OD tolerances ensure 100% compatibility to spectrometers
  • Individually wrapped to prevent contamination
  • Recommended for experiments with small organic molecule (Molecular Weight ∼500) at ambient temperature only
Specifications

Specifications

250 mm
Sample Tube
4 mm
710-SQ-250M
Quartz
5 mm
0.5 mm
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit