Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Multi-tube Jet Solvent NMR/EPR Tube Washer/Dryer
Accommodate up to 5 tubes (3, 4 and 5 mm OD) at once. Bel-Art SP Scienceware™ Wilmad™ Multi-tube Jet Solvent NMR/EPR Tube Washer/Dryer includes calibrated length marks for 4”, 7”, 8” and 9” tubes.
Supplier:  Bel-Art™ 660000066
 View more versions of this product
Catalog No. 16-800-166
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Conveniently provides hands-free operation throughout the washing and drying cycle
  • PTFE solvent cup and PTFE coated transfer tubing eliminate scratches and makes this unit fully resistant to common organic solvents
  • Flanged reservoir connection avoids joint freeze while a greaseless joint between the solvent cup and glass reservoir remains contamination free
  • Disassembles completely without tools for easy cleaning
  • Requires filter flask (sold separately; 1000ml recommended)
Specifications
Specifications
5-tube Washer
Joint type: #9 Silicone Stopper (Top Diameter: 45mm, Bottom Diameter: 37mm, Length: 25mm)
Washer body, solvent cup, and solvent transfer tubing.
25 mm
For tubes 3 to 5mm O.D.
WG-1209-1
Product Suggestions
Product Suggestions