Learn More
Bel-Art™ SP Scienceware™ Wilmad Luer Lock Syringe Needles
Designed for easy sample loading with Luer Lock syringes, these needles are available in stainless steel or PTFE. Bel-Art SP Scienceware™ Wilmad Luer Lock Syringe Needles are ideal for puncturing septum in air-sensitive samples.
Supplier: Bel-Art™ 660000025
Description
- PTFE needles offer a convenient way to wash your small volume tubes by minimizing the possibility of scratches
- Stainless steel needles are designed for easy sample loading
Specifications
1.02 mm | |
Ideal for puncturing septum in air-sensitive samples. | |
12 in. | |
90624 |
0.56 mm | |
Barrel and piston | |
PTFE |