Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Shigemi™ Susceptibility Matched NMR Tubes
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Shigemi™ Susceptibility Matched NMR Tubes feature a susceptibility matched bottom and plunger to effectively reduce the boundary gradients at the edges of the Rf coil reducing the required sample volume by two thirds.
Supplier:  Bel-Art™ SP Scienceware™ 660000032
 View more versions of this product
Catalog No. 16-800-132
Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
  • Select tube based on solvent used: CDCL3, CD3OD, D2O, DMSO-d6
  • Available for Bruker or Universal
Order Info
`
Specifications
Specifications
10 mm (Bottom Length)
Universal, All Spectometers
DMSO-d6 Matched
Tube Set
3 mm
DMS-003