Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Screw-Cap NMR Tube
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Screw-Cap NMR Tube is commonly used in sample degasification due to its capability to maintain vacuum quality.
Supplier:  Bel-Art™ SP Scienceware™ 663050571
 View more versions of this product
Catalog No. 16-800-333
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Each Screw-Cap Tube comes complete with open top screw cap and PTFE/Silicone septum
  • Inert to most organic solvents and compounds but poor to strongly corrosive materials
  • Remains reliable after multiple punctures
Specifications
Specifications
177.8 mm
300 MHz
507-TR-7
5 mm
PTFE/Silicon Septum
Product Suggestions
Product Suggestions