Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Coaxial Small Volume NMR Tubes

Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Coaxial Small Volume NMR Tubes feature a tube within a tube design that allows for easy external referencing.

Supplier:  Bel-Art™ SP Scienceware™ 660000006

 View more versions of this product

Catalog No. 16-800-106

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

  • In complete sets, the tubes are fused together at the bottom
  • Recommended for high field experiments under 600 MHz
  • Tight ID and OD tolerance as small as 0.0065 mm
  • Separately wrapped spacer should replace cap during experiments for improved stability
  • Three unique sample ratios are available for a wide range of applications
  • Complete sets come with inner and outer tubes and cushioning cap for protection during transportation
Specifications

Specifications

178 mm (Outer Tube), 190 mm (Inner Tube)
5 mm outer tube, 2.52 mm inner tube
< 600 MHz
7 in. outer tube , 7.5 in. inner tube
Borosilicate Glass
4.2 mm outer tube, 1.5 mm inner tube
Inner tube, outer tube, spacer, cap with hole
519-Complete
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit