Learn More
Cole-Parmer Wildco 1520-C20 Kemmerer Water Sampler, acrylic, 2.2 L, 3 to 600 ft depth
Supplier: Cole-Parmer 0548610
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Sample at specific depths between 3 and 600 feet. The messenger activates the unique trip heads that ensure closure in fast flowing streams or turbulent waters, regardless of line angle. All models include a plastic carrying case; order messenger and line separately.