Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ UniStore™ Reagent Bottles, Wide Mouth, HDPE, Bulk Packed
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
Quantity:
125 Pk.
250 Pk.
50 Pk.
500 Pk.
Description
UniStore™ Wide Mouth, HDPE, reagent bottles are manufactured from USP Class VI resin that is FDA-21CFR compliant.
- Excellent chemical resistance to most acids, bases, and alcohols
- Polypropylene caps are included
- Manufactured from USP Class VI resin that is FDA-21CFR compliant for food contact and free of heavy metals
- Leakproof closure with integrated seal ring
- Precision molded for consistent wall thickness with smooth interior and exterior surfaces
- Certificates of Analysis for leakproof test details are available upon request
- The 4000 mL bottle is leakproof at ambient temperature and pressure
Specifications
Capacity | 125 mL (4.23 oz.) |
Through the Closure Dispensing | No |
Product Type | Reagent Bottle |
Height (English) | 4 in. |
Closure Material | Polypropylene |
Closure Size | 38 mm |
Height (Metric) | 100 mm |
Capacity (Metric) | 125 mL |
Capacity (English) | 4 oz. |
Color | Translucent |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction