Learn More
Charles River Labs Cell Solutions Whole Blood, 450 mL, Fresh, Kit
Supplier: Charles River Labs Cell Solutions PB000F1KIT
Whole blood is collected from IRB consented healthy human donors in our FDA-registered collection centers. Unless otherwise specified, whole blood is collected in a CPD collection bag and specimen samples are collected in EDTA tubes. We can collect whole blood in any commercially available collection tubes with anticoagulants per your specifications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.