Learn More
Electronic Imaging Materials Inc 667 Vial CryoLabel 1.30 inch x 0.37 inch, 10K per roll, 3 inch core, White
Supplier: Electronic Imaging Materials Inc 667130037
1.30 inch x 0.37 inch CryoLabel will handle extremely cold storage temperatures found in genetics, virology, microbiology and related fields. Conformable—works well on tight diameter test tubes. Resistant to moisture, smudging, and tearing, plus it holds up to alcohol and other chemicals. Designed for thermal transfer and durable UV inkjet printing, allowing you to color-code labels. Scratch and smear resistant when printed with T68 premium resin ribbon.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.