Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Socorex Valve Assembly Parts Kit for Self-Refilling Syringes
Everything needed for valve assembly in one kit.
Specifications
Includes | 2 PTFE washers, 2 corrosion resistant springs, 2 stainless steel balls, Viton™ washer for the barrel |
---|---|
Product Type | Valve Assembly Kit |
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
13-708-24
|
DWK Life Sciences
851426 |
Self-refilling Syringe (2 mL) |
Case for $91.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
13-708-25
|
DWK Life Sciences
851427 |
Self-refilling Syringe (5 mL) |
Case for $42.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
13-708-26
|
DWK Life Sciences
851428 |
Self-refilling Syringe (10 mL) |
Case for $40.25
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Specifications
2 PTFE washers, 2 corrosion resistant springs, 2 stainless steel balls, Viton™ washer for the barrel | |
Valve Assembly Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title