Learn More
Bioworld Wheat Callus Induction Medium (CM4C), 50 L
Supplier: Bioworld 306301214
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Wheat Callus Induction Medium (CM4C)Callus induction medium for monocotsContains MS basal salts with vitamins, sucrose, agar and supplemented with specific growth regulatorsProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested