Learn More
Cytiva Whatman™ Vacuum-Type Glass Membrane Holder
Produced from borosilicate glass and available with a choice of support screen. Whatman™ Vacuum-Type Glass Membrane Holder is suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.
$582.00 - $2469.00
Specifications
Material | Borosilicate Glass |
---|
Includes: Borosilicate glass funnel and base; anodized aluminum clamp; support screen (sintered glass); rubber stopper (No. 8 to fit 1000mL flask).
Catalog Number | Mfr. No. | Volume (Metric) | Includes | Filtration Area | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Includes | Filtration Area | Price | Quantity & Availability | |||
09-907-2
|
Cytiva
1960004 |
300 mL | Prefilter Membrane: 35mm dia. | 9.6 cm2 |
Each for $876.00
|
|
|||
09-907-3
|
Cytiva
1960009 |
1 L | Prefilter Membrane: 70mm dia. | 38.5 cm2 |
Each for $2,469.00
|
|
|||
09-924-548
|
Cytiva
1960002 |
25 mL | Prefilter Membrane: 16 mm dia. | 25 mm |
Pack for $582.00
|
|
|||
Description
Produced from borosilicate glass and available with a choice of support screen. Whatman™ Vacuum-Type Glass Membrane Holder is suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.
- Made from borosilicate glass
- Ensures secure clamping and integral sealing at membrane edge
- Gravimetric analysis filtration
- Ultraclean filtration
- Bacteriological control of food, drink, pharmaceuticals, cosmetics
- Removal of particulates in solution
- Nucleic acid and protein analysis
- HPLC solvent filtration
Specifications
Borosilicate Glass |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.