Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ Swin-Lok Plastic Filter Holder
Designed for microfiltration and ultra cleaning of small volumes of liquids using positive pressure
Supplier: Cytiva 420200
Description
Designed for microfiltration and ultra cleaning of small volumes of liquids using positive pressure
- Syringe compatible
- Accommodates Nuclepore™ track-etched and cast membranes
- Polypropylene
- Membrane: 25mm; Prefilter: 22mm; Filtration Area: 3.9sq.cm
- Female luer-lok cap with male luer slip-fit base
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Swin-Lok Plastic Filter Holder | |
Polypropylene | |
3.9 cm2 | |
3.7 cm |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction