Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ PZ 001 Tweezers
PZ 001 Tweezers are stainless steel forceps with smooth angled jaws (104 mm long) that are highly suitable for handling membrane filters.
Supplier: Cytiva 10477602
Description
- Stainless steel vacuum filtration apparatus accessory
- Autoclavable and can be flame-sterilized with ethanol
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Autoclavable | |
Forceps, smooth angle jaws | |
Stainless Steel | |
104 mm |
Silver Metal | |
Membrane filters in vacuum filtration aparatus | |
1Each | |
8 mm |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction