Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ Puradisc PVDF Syringe Filters: Nonsterile
Designed for high tensile strength, high solvent resistance, and low protein binding. Cytiva Whatman™ Puradisc PVDF Syringe Filters: Nonsterile are well-suited for biomedical filtration, sterilization filtration, and HPLC sample preparation.
Supplier: Cytiva 67492504
Description
- Manufactured from Polyvinylidene Difluoride (PVDF)
- Autoclavable
- Nonsterile
- Includes Female Luer Lock inlet and Male Luer outlet
Specifications
Puradisc 25 mm Polyvinylidene Difluoride Syringe Filter, 0.45 μm (1000 pcs) | |
PVDF | |
Non-sterile | |
HPLC aqueous sample preparation, Biological sample preparation, Buffer solutions, Salt solutions, Tissue culture media, Irrigation solutions, Sterile isolation . |
|
Female luer lock | |
1000Each | |
1000 Pieces | |
Puradisc Syringeless Filter | |
22.9 x 28.4 mm |
PVDF | |
0.45 μm | |
4.2 cm2 | |
Pigment Free Polypropylene (PP) | |
Male luer | |
5.2 bar (75 psi) | |
131°C | |
Up to 100 mL | |
25 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction