Learn More
Cytiva Whatman™ Pop-Top Plastic Filter Holder
Filter holders designed for microfiltration and ultra cleaning of small volumes of liquids using positive pressure.
Supplier: Cytiva 420100
Description
- Designed for microfiltration and ultra cleaning of small volumes of liquids using positive pressure.
- All three holders will accommodate Nuclepore track-etched and cast membranes.
- Syringe compatible.
Specifications
Pop-Top Plastic Filter Holder | |
Polypropylene | |
3.7 cm | |
13 mm |