Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ 0.8 μm Pore Size (ME 27) Mixed Cellulose Ester Membranes, Plain
Whatman™ (ME 27) Mixed Cellulose Ester Membranes, Plain are particularly effective in applications requiring higher flow rates and larger volume filtration including clarification or sterilization of aqueous solutions, particulate analysis and removal, air monitoring, and microbial analysis.
$172.30 - $553.50
Specifications
Autoclavable | Autoclavable |
---|---|
Bubble Point | 1.3 bar |
Filter Type | Plain |
Material | MCE |
Material (Membrane) | ME 27 |
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity & Availability | |||||
09-927-187
|
Cytiva
10400921 |
100 mm |
Pack of 50 for $553.50
|
|
|||||
09-927-186
|
Cytiva
10400906 |
25 mm |
Pack of 100 for $172.30
|
|
|||||
09-927-256
|
Cytiva
10400914 |
50 mm |
Pack of 100 for $197.50
|
|
|||||
Description
- Material: Mixed Cellulose Ester
- Material (Membrane): ME 27
- Filter Type: Plain
- Pore Size: 0.8 μm
- Autoclavable: Yes
- Sterility: Non-sterile
- Bubble Point: 1.3 bar
- Flow Rate: 2.8 sec./100 mL/12.5 cm² (at Δp = 0.9 bar)
- Gas Flow Rate: 11.6 sec./100 mL (at Δp = 3 mbar)
- For Use With (Application): Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Specifications
Autoclavable | |
Plain | |
ME 27 | |
Non-sterile | |
Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays | |
Plain |
1.3 bar | |
MCE | |
0.8 μm | |
2.8 sec./100 mL/12.5 cm⊃ (at Δp = 0.9 bar) | |
11.6 sec./100 mL (at Δp = 3 mbar) |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title