Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ 0.6 μm Pore Size (ME 26) Mixed Cellulose Ester Membranes, Plain
Whatman™ (ME 26) Mixed Cellulose Ester Membranes are particularly effective in applications requiring higher flow rates and larger volume filtration, including clarification or sterilization of aqueous solutions, particulate analysis and removal, air monitoring, and microbial analysis.
$124.00 - $242.00
Specifications
Autoclavable | Autoclavable |
---|---|
Bubble Point | 3.7 bar |
Disposable | Single-use |
Filter Type | Plain |
Material | MCE |
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity & Availability | |||||
09-927-262
|
Cytiva
10401514 |
50 mm |
Pack of 100 for $242.00
|
|
|||||
09-927-831
|
Cytiva
10401506 |
25 mm |
Pack of 100 for $124.00
|
|
|||||
Description
- Filter Type: Plain
- Material: Mixed Cellulose Ester
- Material (Membrane): ME 26
- Pore Size: 0.6 μm
- Autoclavable: Yes
- Sterility: Non-sterile
- Flow Rate: 4.8 sec./100 mL/12.5 cm² (at Δp = 0.9 bar)
- Gas Flow Rate: 21 sec./100 mL (at Δp = 3 mbar)
- Bubble Point: 3.7 bar
- For Use With (Application): Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays
Specifications
Autoclavable | |
Single-use | |
MCE | |
0.6 μm | |
4.8 sec./100 mL/12.5cm² (at Δp = 0.9 bar) | |
21 sec./100 mL (at Δp = 3 mbar) |
3.7 bar | |
Plain | |
ME 26 | |
Non-sterile | |
Filtration, cytology, air monitoring, aqueous HPLC samples, biological assays | |
Plain |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title