Learn More
ZORO TOOLS WESTWARD DIGITAL MULTIMETER
Supplier: ZORO TOOLS G5570966
600 Max. AC Volts, 600 Max. DC Volts, 10 Max. AC Amps, 10 Max. DC Amps. Instrument Counts: 2000Basic DC Accuracy: +/-0.50% of Reading +2 Digits
This item is not returnable.
View return policy