Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Lightweight Cotton Lisle Inspection Gloves

Catalog Number p-2859100
Click to view available options
Gender:
Men's
Women's
This item is not returnable. View return policy
This item is not returnable. View return policy

Lightweight Cotton Lisle Inspection Gloves feature a form-fitting, two-piece design constructed of 100% cotton to provide comfort and absorb perspiration when used as a glove liner underneath liquid-proof gloves.

  • 100% cotton lisle
  • Form-fitting, two-piece construction
  • Can be worn alone or used as a glove liner
  • Unhemmed cuff
  • Moisture absorbing
Construction Cut & Sewn
Coating Material Uncoated
Color White
Disposable Single-use
Hand Ambidextrous
Material Cotton
Product Type Inspection Glove
For Use With (Application) Electronics Assembly, Industrial Manufacturing
Gender Men's
Glove Cuff Style Plain
Series Y6701
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit