Learn More
Terumo BCT Inc TSCD-II Weld Integrity Test Kit
Supplier: Terumo BCT Inc 62000
For use with the TSCD-II sterile tubing welders. The weld integrity test kit is designed to help you satisfy both quality assurance and good manufacturing practice requirements without the need to ship your welder out for performance testing. Welds are evaluated for alignment, leakage and tensile strength.