Learn More
Cole-Parmer Glass Weighing Funnels, medium - fits 100-250 mL flask, 6/pk
Supplier: Cole-Parmer 3470004
These reusable borosilicate glass weighing funnels are designed to ensure complete sample transfer to volumetric flasks. Funnels are flat on one side so you can set them on a balance. Micro funnel is sized to fit microwell plates.